Product Details
- SNP ID
-
rs1049004
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:76979156 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCTCATCTCCGTCCTCGGGGAGGG[C/T]GATGCCAGCGTGGGACTCTTTGGAA
- Phenotype
-
MIM: 159430
MIM: 604760
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MBP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78844309] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MBP
- Gene Name
- myelin basic protein
- Gene
- ZNF236
- Gene Name
- zinc finger protein 236
There are no transcripts associated with this gene.
View Full Product Details