Product Details

SNP ID
rs7112345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:22627260 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAAATGTACAGGCATCACTAATTC[A/G]TTTATTTATTCTTATATTTAACACC
Phenotype
MIM: 613897 MIM: 602835
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FANCF PubMed Links

Gene Details

Gene
FANCF
Gene Name
Fanconi anemia complementation group F
There are no transcripts associated with this gene.

Gene
GAS2
Gene Name
growth arrest specific 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143830.1 1148 Intron NP_001137302.1
NM_005256.3 1148 Intron NP_005247.1
NM_177553.2 1148 Intron NP_808221.1
XM_011519971.2 1148 Intron XP_011518273.1
XM_011519972.2 1148 UTR 5 XP_011518274.1
XM_011519975.1 1148 Intron XP_011518277.1
XM_011519978.2 1148 Intron XP_011518280.1
XM_017017528.1 1148 Intron XP_016873017.1
XM_017017529.1 1148 Intron XP_016873018.1
XM_017017530.1 1148 Intron XP_016873019.1
XM_017017531.1 1148 Intron XP_016873020.1
XM_017017532.1 1148 Intron XP_016873021.1

View Full Product Details