Product Details
- SNP ID
-
rs8076437
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:81313263 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGCTGGCAGGGGAGGCACAGGAAG[A/G]CCCTGTGGTCCCGGGCTACCAGAAT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LINC00482
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75685592] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LINC00482
- Gene Name
- long intergenic non-protein coding RNA 482
There are no transcripts associated with this gene.
- Gene
- TMEM105
- Gene Name
- transmembrane protein 105
View Full Product Details