Product Details
- SNP ID
-
rs34014441
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:61065695 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATTTTGCAGCTGTAAGAAGCAATA[A/G]GCAAGCTTTCTCAGAAAGCTAAGTA
- Phenotype
-
MIM: 611292
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLVS1
PubMed Links
Gene Details
- Gene
- CLVS1
- Gene Name
- clavesin 1
View Full Product Details