Product Details
- SNP ID
-
rs1039825
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:9114098 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACAAAAATAATAGAAAAGATGGTCT[G/T]AAACAAAAGCTGTTCAGTGCTGCTG
- Phenotype
-
MIM: 600532
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NDUFV2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs57716132] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NDUFV2
- Gene Name
- NADH:ubiquinone oxidoreductase core subunit V2
- Gene
- NDUFV2-AS1
- Gene Name
- NDUFV2 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details