Product Details
- SNP ID
-
rs821000
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
79
- Location
-
Chr.1:16447385 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGCCTGCTTCAGATTCCCTATGGGT[A/G]TGAAAACTTCCTCCCTTCTTTCCTC
- Phenotype
-
MIM: 611624
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NECAP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs72638050] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NECAP2
- Gene Name
- NECAP endocytosis associated 2
- Gene
- SPATA21
- Gene Name
- spermatogenesis associated 21
There are no transcripts associated with this gene.
View Full Product Details