Product Details
- SNP ID
-
rs10986355
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:124486020 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGATACCGCAGTAGGGGCAGGGGC[A/G]GGAGGGGGCTGGGGAAGACCAGGGC
- Phenotype
-
MIM: 184757
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADGRD2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs180948653] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADGRD2
- Gene Name
- adhesion G protein-coupled receptor D2
There are no transcripts associated with this gene.
- Gene
- NR5A1
- Gene Name
- nuclear receptor subfamily 5 group A member 1
View Full Product Details