Product Details
- SNP ID
-
rs28684
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:38873760 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACCAAGCTAGGCTGGTGCCCCGGC[C/G]AGGGTGGGGCTGGCATTGCCTCTGC
- Phenotype
-
MIM: 602920
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LASP1
PubMed Links
Gene Details
- Gene
- LASP1
- Gene Name
- LIM and SH3 protein 1
View Full Product Details