Product Details

SNP ID
rs6801699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:129556776 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCAAAGCCAGCTGCTTGCACACAA[C/T]CCCCAACTCCCCACGGCCCCCACAG
Phenotype
MIM: 604282
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
H1FOO PubMed Links

Gene Details

Gene
H1FOO
Gene Name
H1 histone family member O, oocyte specific
There are no transcripts associated with this gene.

Gene
PLXND1
Gene Name
plexin D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015103.2 Intron NP_055918.2
XM_011512588.1 Intron XP_011510890.1
XM_011512589.1 Intron XP_011510891.1
XM_011512590.2 Intron XP_011510892.1
XM_011512592.1 Intron XP_011510894.1

View Full Product Details