Product Details
- SNP ID
-
rs2255655
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:12557340 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGAGGCAGAGCGAGCGCTGCCGCCA[C/G]AGCCGGGGATCCGGGGGATGCGTGG
- Phenotype
-
MIM: 608426
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MKRN2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112706793] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MKRN2
- Gene Name
- makorin ring finger protein 2
- Gene
- MKRN2OS
- Gene Name
- MKRN2 opposite strand
View Full Product Details