Product Details

SNP ID
rs17645143
Assay Type
Functionally tested
NCBI dbSNP Submissions
53
Location
Chr.1:109293135 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTTTTTACCATGCTCATGATCC[C/T]GAGGCGTTTACTCAGTCAGCAGGAG
Phenotype
MIM: 613126
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYBPHL PubMed Links

Gene Details

Gene
MYBPHL
Gene Name
myosin binding protein H like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010985.2 Intron NP_001010985.2
NM_001265613.1 Intron NP_001252542.1
XM_017001173.1 Intron XP_016856662.1
XM_017001174.1 Intron XP_016856663.1
XM_017001175.1 Intron XP_016856664.1
Gene
PSRC1
Gene Name
proline and serine rich coiled-coil 1
There are no transcripts associated with this gene.

View Full Product Details