Product Details
- SNP ID
-
rs71575359
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:111093966 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTAAATGTTCTTTACCCAGACTGTA[C/G]ACTGCTCAGATTAACCAATGCATTG
- Phenotype
-
MIM: 609669
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
WDR36
PubMed Links
Gene Details
- Gene
- WDR36
- Gene Name
- WD repeat domain 36
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_139281.2 |
|
Intron |
|
|
NP_644810.1 |
View Full Product Details