Product Details

SNP ID
rs3792879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132309142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGCCGGTTTCCTAAGAAAAGCC[C/T]TTTCTAAAGGACCCCTCTTAGGACT
Phenotype
MIM: 604190
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC553103 PubMed Links

Gene Details

Gene
LOC553103
Gene Name
uncharacterized LOC553103
There are no transcripts associated with this gene.

Gene
SLC22A4
Gene Name
solute carrier family 22 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003059.2 Intron NP_003050.2
XM_006714675.3 Intron XP_006714738.1
XM_011543589.2 Intron XP_011541891.1
XM_017009776.1 Intron XP_016865265.1

View Full Product Details