Product Details

SNP ID
rs12401504
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:22644736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGTTGCTGTGAGGATTAAGTTA[A/G]CTTAAATATAAAACAGCCCTGAGCA
Phenotype
MIM: 120550 MIM: 120570 MIM: 120575
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1QA PubMed Links

Gene Details

Gene
C1QA
Gene Name
complement C1q A chain
There are no transcripts associated with this gene.

Gene
C1QB
Gene Name
complement C1q B chain
There are no transcripts associated with this gene.

Gene
C1QC
Gene Name
complement C1q C chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114101.1 Intron NP_001107573.1
NM_172369.3 Intron NP_758957.2

View Full Product Details