Product Details
- SNP ID
-
rs2649655
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:57554047 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTAGCACAGTGCCGAAAGTGCTC[A/G]GTCAAGTGACAGGTATTATCATTAT
- Phenotype
-
MIM: 613664
MIM: 603890
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SMTNL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74563249] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SMTNL1
- Gene Name
- smoothelin like 1
There are no transcripts associated with this gene.
- Gene
- UBE2L6
- Gene Name
- ubiquitin conjugating enzyme E2 L6
View Full Product Details