Product Details

SNP ID
rs11832131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55956052 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGGTGTGCTTCCACTGACCAGCC[A/C]TAGGTAGCACACAGCTGCCTCCATC
Phenotype
MIM: 125855 MIM: 155550
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DGKA PubMed Links

Gene Details

Gene
DGKA
Gene Name
diacylglycerol kinase alpha
There are no transcripts associated with this gene.

Gene
PMEL
Gene Name
premelanosome protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001200053.1 Intron NP_001186982.1
NM_001200054.1 Intron NP_001186983.1
NM_001320121.1 Intron NP_001307050.1
NM_001320122.1 Intron NP_001307051.1
NM_006928.4 Intron NP_008859.1
XM_006719569.1 Intron XP_006719632.1
XM_011538685.1 Intron XP_011536987.1

View Full Product Details