Product Details
- SNP ID
-
rs71382566
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:18794701 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGAAGTACTAGCAATGAAAACAAG[A/G]ATTTAATATCAAAACTTCATGTGAC
- Phenotype
-
MIM: 607669
MIM: 603674
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ARL6IP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113687501] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARL6IP1
- Gene Name
- ADP ribosylation factor like GTPase 6 interacting protein 1
- Gene
- RPS15A
- Gene Name
- ribosomal protein S15a
There are no transcripts associated with this gene.
View Full Product Details