Product Details

SNP ID
rs2606558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:18809869 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAGAGAGCTAAGACAGTTATAAA[C/G]ATATGAAAAGCACACAATCCATTAA
Phenotype
MIM: 607669 MIM: 607032
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARL6IP1 PubMed Links

Gene Details

Gene
ARL6IP1
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 1
There are no transcripts associated with this gene.

Gene
SMG1
Gene Name
SMG1, nonsense mediated mRNA decay associated PI3K related kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015092.4 Intron NP_055907.3
XM_005255182.3 Intron XP_005255239.1
XM_005255183.3 Intron XP_005255240.1
XM_005255184.3 Intron XP_005255241.1
XM_011545769.2 Intron XP_011544071.1
XM_011545770.1 Intron XP_011544072.1
XM_017023066.1 Intron XP_016878555.1
XM_017023067.1 Intron XP_016878556.1

View Full Product Details