Product Details

SNP ID
rs7555175
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:160797378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATATGGGTGAACGAGGAGCCTAGA[A/C]CTCAGAGCTGAGGTCCAGTCTGACC
Phenotype
MIM: 600684
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC105371470 PubMed Links

Gene Details

Gene
LOC105371470
Gene Name
uncharacterized LOC105371470
There are no transcripts associated with this gene.

Gene
LY9
Gene Name
lymphocyte antigen 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033667.2 Intron NP_001028839.1
NM_001261456.1 Intron NP_001248385.1
NM_001261457.1 Intron NP_001248386.1
NM_002348.3 Intron NP_002339.2
XM_011509548.1 Intron XP_011507850.1
XM_011509549.1 Intron XP_011507851.1
XM_011509550.1 Intron XP_011507852.1
XM_011509552.1 Intron XP_011507854.1
XM_011509556.1 Intron XP_011507858.1
XM_011509560.1 Intron XP_011507862.1
XM_017001297.1 Intron XP_016856786.1
XM_017001298.1 Intron XP_016856787.1
XM_017001299.1 Intron XP_016856788.1
XM_017001300.1 Intron XP_016856789.1
XM_017001301.1 Intron XP_016856790.1
XM_017001302.1 Intron XP_016856791.1
XM_017001303.1 Intron XP_016856792.1
XM_017001304.1 Intron XP_016856793.1

View Full Product Details