Product Details

SNP ID
rs3742130
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:99255087 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAGGCTCCCCAGGGATTGTAACT[A/G]TTCTCCCCCGTTCCCAGCATCAGGA
Phenotype
MIM: 602042
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR18 PubMed Links

Gene Details

Gene
GPR18
Gene Name
G protein-coupled receptor 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098200.1 1287 Silent Mutation AAC,AAT N262N NP_001091670.1
NM_005292.3 1287 Silent Mutation AAC,AAT N262N NP_005283.1
XM_006719946.3 1287 Silent Mutation AAC,AAT N262N XP_006720009.1
Gene
UBAC2
Gene Name
UBA domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144072.1 1287 Intron NP_001137544.1
NM_177967.3 1287 Intron NP_808882.1
XM_006719948.3 1287 Intron XP_006720011.1
XM_011521082.2 1287 Intron XP_011519384.1
XM_011521083.2 1287 Intron XP_011519385.1
XM_011521084.2 1287 Intron XP_011519386.1
XM_017020553.1 1287 Intron XP_016876042.1

View Full Product Details