Product Details

SNP ID
rs3780297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:77727686 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTGTCAGGCATCATGCTAAACGTA[C/T]ACATATGTTATATACTACAGTGCAC
Phenotype
MIM: 600998
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNAQ PubMed Links

Gene Details

Gene
GNAQ
Gene Name
G protein subunit alpha q
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002072.4 Intron NP_002063.2
XM_017014628.1 Intron XP_016870117.1

View Full Product Details