Product Details

SNP ID
rs12708952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:55515948 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGACATAGAGAAATTTAAAAAGAC[C/G]TAAGACATACAGAAAACAAATAGCA
Phenotype
MIM: 612040 MIM: 120360
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LPCAT2 PubMed Links

Gene Details

Gene
LPCAT2
Gene Name
lysophosphatidylcholine acyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017839.4 Intron NP_060309.2
XM_005256006.2 Intron XP_005256063.1
XM_011523169.2 Intron XP_011521471.1
XM_017023376.1 Intron XP_016878865.1
Gene
MMP2
Gene Name
matrix metallopeptidase 2
There are no transcripts associated with this gene.

View Full Product Details