Product Details
- SNP ID
-
rs11639329
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:55540218 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTACCAAGTAAACAGCATTCAAGT[C/T]CTTGAATCAATGGCTAAAAATTCCA
- Phenotype
-
MIM: 606902
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC107984745
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117471766] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC107984745
- Gene Name
- uncharacterized mitochondrial protein AtMg00860-like
There are no transcripts associated with this gene.
- Gene
- PYGO1
- Gene Name
- pygopus family PHD finger 1
View Full Product Details