Product Details

SNP ID
rs707916
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:31729781 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCTGGCTGGTCACGCTGCCCCTAGC[A/G]CGACCTAGTATAAACCAGACCGAGT
Phenotype
MIM: 606520 MIM: 602872 MIM: 604744 MIM: 610435
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C6orf25 PubMed Links

Gene Details

Gene
C6orf25
Gene Name
chromosome 6 open reading frame 25
There are no transcripts associated with this gene.

Gene
CLIC1
Gene Name
chloride intracellular channel 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287593.1 485 Intron NP_001274522.1
NM_001287594.1 485 Intron NP_001274523.1
NM_001288.4 485 Intron NP_001279.2
Gene
DDAH2
Gene Name
dimethylarginine dimethylaminohydrolase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303007.1 485 UTR 5 NP_001289936.1
NM_001303008.1 485 Intron NP_001289937.1
NM_013974.2 485 Intron NP_039268.1
XM_011514448.2 485 UTR 5 XP_011512750.1
Gene
LY6G6C
Gene Name
lymphocyte antigen 6 complex, locus G6C
There are no transcripts associated with this gene.

View Full Product Details