Product Details
- SNP ID
-
rs453098
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:31723880 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGCTGGAGCTCCTCTTGAGCGCGGG[A/G]GACTCGGGCACTTTTTTCTGCAAGG
- Phenotype
-
MIM: 606520
MIM: 602872
MIM: 604744
MIM: 610435
MIM: 606038
MIM: 610437
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C6orf25
PubMed Links
Gene Details
- Gene
- C6orf25
- Gene Name
- chromosome 6 open reading frame 25
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_025260.3 |
628 |
Silent Mutation |
GGA,GGG |
G101G |
NP_079536.2 |
NM_138272.2 |
628 |
Silent Mutation |
GGA,GGG |
G101G |
NP_612116.1 |
NM_138273.2 |
628 |
Silent Mutation |
GGA,GGG |
G101G |
NP_612117.1 |
NM_138274.2 |
628 |
Silent Mutation |
GGA,GGG |
G101G |
NP_612118.1 |
NM_138275.2 |
628 |
Silent Mutation |
GGA,GGG |
G101G |
NP_612119.1 |
NM_138277.2 |
628 |
Silent Mutation |
GGA,GGG |
G101G |
NP_612121.1 |
XM_011514920.2 |
628 |
Silent Mutation |
GGA,GGG |
G117G |
XP_011513222.1 |
XM_011514921.2 |
628 |
Silent Mutation |
GGA,GGG |
G117G |
XP_011513223.1 |
XM_017011330.1 |
628 |
Silent Mutation |
GGA,GGG |
G117G |
XP_016866819.1 |
XM_017011331.1 |
628 |
Silent Mutation |
GGA,GGG |
G101G |
XP_016866820.1 |
XM_017011332.1 |
628 |
Silent Mutation |
GGA,GGG |
G117G |
XP_016866821.1 |
XM_017011333.1 |
628 |
Silent Mutation |
GGA,GGG |
G117G |
XP_016866822.1 |
- Gene
- CLIC1
- Gene Name
- chloride intracellular channel 1
There are no transcripts associated with this gene.
- Gene
- DDAH2
- Gene Name
- dimethylarginine dimethylaminohydrolase 2
There are no transcripts associated with this gene.
- Gene
- LY6G6C
- Gene Name
- lymphocyte antigen 6 complex, locus G6C
There are no transcripts associated with this gene.
- Gene
- LY6G6D
- Gene Name
- lymphocyte antigen 6 complex, locus G6D
There are no transcripts associated with this gene.
- Gene
- LY6G6E
- Gene Name
- lymphocyte antigen 6 complex, locus G6E (pseudogene)
There are no transcripts associated with this gene.
View Full Product Details