Product Details

SNP ID
rs2845702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:120242952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGGAAAACAGTGGGAAGCCAGC[G/T]GGCGGGGGCACACCTGGTTTGCCCC
Phenotype
MIM: 607394
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC649133 PubMed Links

Gene Details

Gene
LOC649133
Gene Name
uncharacterized LOC649133
There are no transcripts associated with this gene.

Gene
POU2F3
Gene Name
POU class 2 homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244682.1 2258 Intron NP_001231611.1
NM_014352.3 2258 Intron NP_055167.2
XM_011542739.2 2258 Intron XP_011541041.1
XM_011542740.2 2258 Intron XP_011541042.1
XM_011542741.2 2258 Intron XP_011541043.1
XM_011542742.2 2258 Intron XP_011541044.1
XM_011542743.2 2258 UTR 5 XP_011541045.1
XM_017017487.1 2258 Intron XP_016872976.1

View Full Product Details