Product Details

SNP ID
rs12622908
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:121346662 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAAGGTGAGAGCCAGCTGTTGCCAC[C/T]ACTGATGCCCATGAGGGACTCAGCT
Phenotype
MIM: 605852
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLASP1 PubMed Links

Gene Details

Gene
CLASP1
Gene Name
cytoplasmic linker associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142273.1 Intron NP_001135745.1
NM_001142274.1 Intron NP_001135746.1
NM_001207051.1 Intron NP_001193980.1
NM_015282.2 Intron NP_056097.1
XM_006712377.1 Intron XP_006712440.1
XM_006712381.1 Intron XP_006712444.1
XM_006712382.1 Intron XP_006712445.1
XM_006712383.1 Intron XP_006712446.1
XM_006712385.1 Intron XP_006712448.1
XM_011510848.1 Intron XP_011509150.1
XM_011510849.1 Intron XP_011509151.1
XM_017003657.1 Intron XP_016859146.1
XM_017003658.1 Intron XP_016859147.1
XM_017003659.1 Intron XP_016859148.1
XM_017003660.1 Intron XP_016859149.1
XM_017003661.1 Intron XP_016859150.1
XM_017003662.1 Intron XP_016859151.1
XM_017003663.1 Intron XP_016859152.1
XM_017003664.1 Intron XP_016859153.1
XM_017003665.1 Intron XP_016859154.1
XM_017003666.1 Intron XP_016859155.1
XM_017003667.1 Intron XP_016859156.1
XM_017003668.1 Intron XP_016859157.1
XM_017003669.1 Intron XP_016859158.1
XM_017003670.1 Intron XP_016859159.1
XM_017003671.1 Intron XP_016859160.1
XM_017003672.1 Intron XP_016859161.1
XM_017003673.1 Intron XP_016859162.1
XM_017003674.1 Intron XP_016859163.1
XM_017003675.1 Intron XP_016859164.1
XM_017003676.1 Intron XP_016859165.1
XM_017003677.1 Intron XP_016859166.1
XM_017003678.1 Intron XP_016859167.1
XM_017003679.1 Intron XP_016859168.1
XM_017003680.1 Intron XP_016859169.1
XM_017003681.1 Intron XP_016859170.1
XM_017003682.1 Intron XP_016859171.1
XM_017003683.1 Intron XP_016859172.1
XM_017003684.1 Intron XP_016859173.1
XM_017003685.1 Intron XP_016859174.1
XM_017003686.1 Intron XP_016859175.1
XM_017003687.1 Intron XP_016859176.1
XM_017003688.1 Intron XP_016859177.1
XM_017003689.1 Intron XP_016859178.1
XM_017003690.1 Intron XP_016859179.1
XM_017003691.1 Intron XP_016859180.1

View Full Product Details