Product Details

SNP ID
rs3200401
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:65504361 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAATGCAGTTGTCTTGACTTCAGGT[T/C]TGTCTGTTCTGTTGGCAAGTAAATG
Phenotype
MIM: 607924
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
MALAT1 PubMed Links

Gene Details

Gene
MALAT1
Gene Name
metastasis associated lung adenocarcinoma transcript 1 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details