Product Details

SNP ID
rs1328866
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:8653283 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAACACAAGGCAGAGAATAAGATT[A/T]GAGTAAAAGAGAGAGTTATATAAAA
Phenotype
MIM: 612210
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HULC PubMed Links

Gene Details

Gene
HULC
Gene Name
hepatocellular carcinoma up-regulated long non-coding RNA
There are no transcripts associated with this gene.

Gene
LOC100506207
Gene Name
uncharacterized LOC100506207
There are no transcripts associated with this gene.

View Full Product Details