Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_006898.4 | 2967 | Intron | NP_008829.3 | ||
XM_005246509.3 | 2967 | Intron | XP_005246566.1 | ||
XM_005246510.4 | 2967 | Intron | XP_005246567.1 | ||
XM_005246511.3 | 2967 | Intron | XP_005246568.1 | ||
XM_005246513.4 | 2967 | Intron | XP_005246570.1 | ||
XM_006712477.2 | 2967 | Intron | XP_006712540.1 | ||
XM_011511065.2 | 2967 | Intron | XP_011509367.1 | ||
XM_011511066.2 | 2967 | Intron | XP_011509368.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_014621.2 | 2967 | Intron | NP_055436.2 | ||
XM_005246514.4 | 2967 | UTR 3 | XP_005246571.1 |