Product Details

SNP ID
rs10930722
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:176089768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACAGTGGGACTTTGGGCACCAT[A/G]GGGTCATCCTGAGTTTTTCACCAAA
Phenotype
MIM: 142991 MIM: 142989
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EVX2 PubMed Links

Gene Details

Gene
EVX2
Gene Name
even-skipped homeobox 2
There are no transcripts associated with this gene.

Gene
HOXD13
Gene Name
homeobox D13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000523.3 1331 Intron NP_000514.2
XM_011511068.2 1331 Intron XP_011509370.1
XM_011511069.2 1331 UTR 3 XP_011509371.1

View Full Product Details