Product Details

SNP ID
rs11833550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:9856081 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACAAGCATTCTTCCCTGATACATT[C/T]GAAGATTTCTCTTAAGTTCAAAACT
Phenotype
MIM: 603242
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLEC2B PubMed Links

Gene Details

Gene
CLEC2B
Gene Name
C-type lectin domain family 2 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005127.2 Intron NP_005118.2

View Full Product Details