Product Details

SNP ID
rs10260448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:155502282 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTTAGTAGTAAACTCAACACACT[A/G]AATGTGAGCCAAAAAAAAAACGTTT
Phenotype
MIM: 612493
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNPY1 PubMed Links

Gene Details

Gene
CNPY1
Gene Name
canopy FGF signaling regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001103176.1 1210 UTR 3 NP_001096646.1

View Full Product Details