Product Details

SNP ID
rs36099621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:155505014 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGTTTTTATCATTGATGTGTTG[C/T]TTGAGCTTTTTCTAGCAAAAATTGT
Phenotype
MIM: 612493
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNPY1 PubMed Links

Gene Details

Gene
CNPY1
Gene Name
canopy FGF signaling regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001103176.1 Intron NP_001096646.1

View Full Product Details