Product Details
- SNP ID
-
rs2187118
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:46191396 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TAGGAAAGCAAGTAGTCCATCCCGC[A/G]TCAGTCATGGCACCATCAGCCCAGC
- Phenotype
-
MIM: 600909
MIM: 612412
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LSS
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73144744] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LSS
- Gene Name
- lanosterol synthase (2,3-oxidosqualene-lanosterol cyclase)
- Gene
- SPATC1L
- Gene Name
- spermatogenesis and centriole associated 1-like
There are no transcripts associated with this gene.
View Full Product Details