Product Details

SNP ID
rs2247002
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:28430174 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCGTGTCCTCCGTTCCCTAGGTCCC[C/G]GGTCCTCCAGGGTCTCAGGCATCAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZSCAN23 PubMed Links

Gene Details

Gene
ZSCAN23
Gene Name
zinc finger and SCAN domain containing 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012455.1 3240 Intron NP_001012458.1
XM_005248950.2 3240 UTR 3 XP_005249007.1
XM_011514406.2 3240 Intron XP_011512708.1
XM_011514408.2 3240 Intron XP_011512710.1
XM_011514410.2 3240 Intron XP_011512712.1
XM_017010479.1 3240 Intron XP_016865968.1

View Full Product Details