Product Details

SNP ID
rs2069885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:135892476 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAAATGTCAGCGCGTTGCCTGCC[G/A]TGGTTTGGTTGCATGGCTGTTCACA
Phenotype
MIM: 146931 MIM: 616150
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
IL9 PubMed Links

Gene Details

Gene
IL9
Gene Name
interleukin 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000590.1 361 Missense Mutation ACG,ATG T117M NP_000581.1
Gene
SLC25A48
Gene Name
solute carrier family 25 member 48
There are no transcripts associated with this gene.

View Full Product Details