Product Details

SNP ID
rs4796137
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:35924448 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GGTATTGCCTACAGTAAAACCCTAT[A/G]TTGTAAGACAGAAAAACTCTGCCCA
Phenotype
MIM: 612896
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RDM1 PubMed Links

Gene Details

Gene
RDM1
Gene Name
RAD52 motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034836.1 Intron NP_001030008.1
NM_001163120.1 Intron NP_001156592.1
NM_001163121.1 Intron NP_001156593.1
NM_001163122.1 Intron NP_001156594.1
NM_001163124.1 Intron NP_001156596.1
NM_001163125.1 Intron NP_001156597.1
NM_001163130.1 Intron NP_001156602.1
NM_145654.3 Intron NP_663629.1
XM_011524509.2 Intron XP_011522811.1
XM_011524510.2 Intron XP_011522812.1

View Full Product Details