Product Details
- SNP ID
-
rs2413033
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:31178227 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GTTCCAGTTGGACTAAATGCTGATT[A/G]ATCTGAATTCAGCAGATTTGTCCCT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RNF185
PubMed Links
Gene Details
- Gene
- RNF185
- Gene Name
- ring finger protein 185
View Full Product Details