Product Details
- SNP ID
-
rs2537522
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:67320042 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGATGTGTGCTCTACGGGTTCTTA[G/T]AATAGCAATGAACCTTCATAATCAG
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
STAG3L4
PubMed Links
Gene Details
- Gene
- STAG3L4
- Gene Name
- stromal antigen 3-like 4 (pseudogene)
There are no transcripts associated with this gene.
View Full Product Details