Product Details

SNP ID
rs11740219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:54978323 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACCCTCTGTTGCTCATTTTTTGA[C/T]ATTTTTGAAATCCAGAGTGACTCTG
Phenotype
MIM: 601521
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESM1 PubMed Links

Gene Details

Gene
ESM1
Gene Name
endothelial cell specific molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135604.1 1633 UTR 3 NP_001129076.1
NM_007036.4 1633 UTR 3 NP_008967.1

View Full Product Details