Product Details

SNP ID
rs11953294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:135579610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTCAAAGAGGGTATGTGTGTGT[A/G]TGCGGGGGTGGGGACGGGTATTGGG
Phenotype
MIM: 604186 MIM: 616150
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXCL14 PubMed Links

Gene Details

Gene
CXCL14
Gene Name
C-X-C motif chemokine ligand 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004887.4 Intron NP_004878.2
Gene
SLC25A48
Gene Name
solute carrier family 25 member 48
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145282.4 Intron NP_660325.4
XM_005271903.1 Intron XP_005271960.1
XM_006714544.1 Intron XP_006714607.1
XM_006714546.1 Intron XP_006714609.1
XM_006714550.1 Intron XP_006714613.1
XM_006714552.2 Intron XP_006714615.1
XM_011543196.1 Intron XP_011541498.1
XM_011543199.1 Intron XP_011541501.1
XM_017009086.1 Intron XP_016864575.1
XM_017009087.1 Intron XP_016864576.1
XM_017009088.1 Intron XP_016864577.1
XM_017009089.1 Intron XP_016864578.1
XM_017009090.1 Intron XP_016864579.1
XM_017009091.1 Intron XP_016864580.1
XM_017009092.1 Intron XP_016864581.1

View Full Product Details