Product Details

SNP ID
rs8049746
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3194096 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAACTTTGCAAGAATAAGAAGAAA[C/T]GGCATGAATTTGGAAGAAAAACAGG
Phenotype
MIM: 603232
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR1F1 PubMed Links

Gene Details

Gene
OR1F1
Gene Name
olfactory receptor family 1 subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012360.1 Intron NP_036492.1
XM_011522506.2 Intron XP_011520808.1
XM_011522507.2 Intron XP_011520809.1

View Full Product Details