Product Details

SNP ID
rs13359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:74448155 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGAGCTCTTCACCCTAAAGATTC[A/G]GTCTGGTTTGCTAATGACTTATCTA
Phenotype
MIM: 600753
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLG1 PubMed Links

Gene Details

Gene
GLG1
Gene Name
golgi glycoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145666.1 7530 UTR 3 NP_001139138.1
NM_001145667.1 7530 UTR 3 NP_001139139.1
NM_012201.5 7530 UTR 3 NP_036333.2

View Full Product Details