Product Details

SNP ID
rs4336593
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142658233 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTACTGCCTGTCTCAGTCAGTTT[G/T]GATTGCGATAACAAAATACCACAGG
Phenotype
MIM: 603210
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
JRK PubMed Links

Gene Details

Gene
JRK
Gene Name
Jrk helix-turn-helix protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077527.2 9453 UTR 3 NP_001070995.2
NM_001279352.1 9453 Intron NP_001266281.1
NM_003724.3 9453 UTR 3 NP_003715.3
XM_006716677.3 9453 UTR 3 XP_006716740.1
XM_006716678.3 9453 UTR 3 XP_006716741.1
XM_011517354.1 9453 Intron XP_011515656.1
XM_011517355.1 9453 Intron XP_011515657.1

View Full Product Details