Product Details

SNP ID
rs13632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:47783455 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTCACATTTCTGTGTGGAGCCTG[A/G]GGCTGTGTGCAAAGTCTGGGGTTTG
Phenotype
MIM: 606542 MIM: 612187
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HDAC7 PubMed Links

Gene Details

Gene
HDAC7
Gene Name
histone deacetylase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098416.3 3458 UTR 3 NP_001091886.1
NM_001308090.1 3458 UTR 3 NP_001295019.1
NM_015401.4 3458 UTR 3 NP_056216.2
XM_011538478.1 3458 UTR 3 XP_011536780.1
XM_011538479.1 3458 UTR 3 XP_011536781.1
XM_011538480.1 3458 UTR 3 XP_011536782.1
XM_011538481.1 3458 UTR 3 XP_011536783.1
XM_011538482.1 3458 UTR 3 XP_011536784.1
XM_011538483.1 3458 Intron XP_011536785.1
XM_017019455.1 3458 UTR 3 XP_016874944.1
XM_017019456.1 3458 Intron XP_016874945.1
Gene
SLC48A1
Gene Name
solute carrier family 48 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017842.2 3458 Intron NP_060312.2
XM_005269016.4 3458 Intron XP_005269073.1
XM_011538558.2 3458 Intron XP_011536860.1
XM_017019612.1 3458 Intron XP_016875101.1
XM_017019613.1 3458 Intron XP_016875102.1
XM_017019614.1 3458 Intron XP_016875103.1
XM_017019615.1 3458 Intron XP_016875104.1
XM_017019616.1 3458 Intron XP_016875105.1
XM_017019617.1 3458 Intron XP_016875106.1

View Full Product Details