Product Details

SNP ID
rs10733735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104751639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAATTCAACCATTTAAGGGACTT[C/T]ACAATATGGAGGTTTATAGGGGATG
Phenotype
MIM: 608871
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NIPSNAP3A PubMed Links

Gene Details

Gene
NIPSNAP3A
Gene Name
nipsnap homolog 3A
There are no transcripts associated with this gene.

View Full Product Details