Product Details

SNP ID
rs6927768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:170557897 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCTTCCCCTTTATAGTCACTCTG[C/T]TGGCCCCAGGTAACTACTGCATCTG
Phenotype
MIM: 602017 MIM: 600075
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMB1 PubMed Links

Gene Details

Gene
PSMB1
Gene Name
proteasome subunit beta 1
There are no transcripts associated with this gene.

Gene
TBP
Gene Name
TATA-box binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172085.1 Intron NP_001165556.1
NM_003194.4 Intron NP_003185.1

View Full Product Details