Product Details

SNP ID
rs10067976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134761600 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTAGAGAAACCTCTTCCTTGAACT[A/C]CAGGCTAATATTTCAACTATATACT
Phenotype
MIM: 601118
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CAMLG PubMed Links

Gene Details

Gene
CAMLG
Gene Name
calcium modulating ligand
There are no transcripts associated with this gene.

Gene
DDX46
Gene Name
DEAD-box helicase 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300860.1 Intron NP_001287789.1
NM_014829.3 Intron NP_055644.2
XM_011543758.1 Intron XP_011542060.1

View Full Product Details