Product Details

SNP ID
rs815595
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:293909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCACTGGAATCCATCAGCACCA[T/C]GAGCCTTCTGCTTTTTAAATTAAAA
Phenotype
MIM: 616778
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
DUSP22 PubMed Links

Gene Details

Gene
DUSP22
Gene Name
dual specificity phosphatase 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286555.1 Intron NP_001273484.1
NM_020185.4 Intron NP_064570.1
XM_011514757.2 Intron XP_011513059.1
XM_017011061.1 Intron XP_016866550.1
XM_017011062.1 Intron XP_016866551.1

View Full Product Details